Would you like to log in again?
OxiSelect™ DNA Double-Strand Break (DSB) Staining Kit, Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™
STA-321 STA-321-T
102964-908EA 363.08 USD InStock
102964-908 102964-906
OxiSelect™ DNA Double-Strand Break (DSB) Staining Kit, Cell Biolabs
Assays Cellular Assays
The OxiSelect™ DNA Double-Strand Break Staining Kit provides an easy-to-use method for detecting the presence of DSBs in cells cultured in microtiter plates
Double-strand breaks (DSB) in DNA are among the most dangerous types of DNA damage occuring within cells. One of the earliest cellular responses to double-strand breaks is the phosphorylation of a histone variant, H2AX, at the sites of DNA damage. With this kit, double strand breaks can be detected in just a few hours by immunofluorescence staining of the phosphorylated histone H2AX
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty