Would you like to log in again?
OxiSelect™ UV-Induced DNA Damage ELISA Kit (6-4PP Quantitation), Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™
STA-323 STA-323-5
102964-920EA 3304.62 USD InStock
102964-920 102964-918
OxiSelect™ UV-Induced DNA Damage ELISA Kit (6-4PP Quantitation), Cell Biolabs
Assays ELISAs
OxiSelect™ UV-Induced DNA Damage ELISA Kit measures 6-4PP formation in DNA isolated from cells or tissue
Standards or unknown DNA samples are first heat denatured before adsorbed onto a 96-well DNA binding plate.  The sample or standard is then probed with an anti-6-4PP antibody, followed by an HRP conjugated secondary antibody.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty