Would you like to log in again?
OxiSelect™ Catalase Activity Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-339 STA-341
102964-970EA 763.08 USD InStock
102964-970 102964-962
OxiSelect™ Catalase Activity Assay Kits, Cell Biolabs
Assays Enzyme Assays
Catalase is a ubiquitous enzyme that destroys hydrogen peroxides formed during oxidative stress
The OxiSelect™ Catalase Activity Assay Kit (Fluorometric) provides a 40-fold increase in sensitivity compared to our colorimetric assay. Each kit provides sufficient reagents to perform up to 500 assays in a 96-well microtiter plate. This includes blanks, catalase standards and unknown samples.
OxiSelect™ Catalase Activity Assay Kits measure catalase activity in less than one hour from a variety of samples including blood, cells and tissues. The colorimetric kit provides sufficient reagents to perform up to 100 assays in a 96-well microtiter plate, or 50 assays in microcentrifuge tubes. This includes blanks, catalase standards and unknown samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty