Would you like to log in again?
OxiSelect™ 96-Well Comet Assay Kits and Slides, Cell Biolabs
Supplier: Cell Biolabs
STA-355-5 STA-356-5 STA-356 STA-355
102964-992EA 455.38 USD InStock
102964-992 102964-994 102964-996 102964-998
OxiSelect™ 96-Well Comet Assay Kits and Slides, Cell Biolabs
Assays Cellular Assays
OxiSelect™ 96-Well Comet Assay Kits and Slides provide a fast, convenient way to screen for general DNA damage, regardless of the source or nature of the damage
Kits include Comet Slides, reagents, and a fluorescent dye to visualize cells under an epifluorescence microscope. Slides are available to order separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty