Would you like to log in again?
Urinary Creatinine Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-378
102965-044EA 467.69 USD InStock
102965-044
Urinary Creatinine Assay Kit, Cell Biolabs
Assays Immunological Assays
Cell Biolabs’ Creatinine Assay Kit measures creatinine levels in urine
Samples are compared to a known concentration of creatinine standard within a 96-well microtiter plate format. Samples and standards are incubated for 30 minutes with a reaction reagent which changes color from yellow to bright orange upon reacting with creatinine, forming the creatinine-picrate complex. The plate is read with a standard 96-well spectrophotometric microplate reader at 490 nm. This kit can detect as little as 300 µg/dL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty