Would you like to log in again?
Global DNA Methylation ELISA Kit (5’-methyl-2’-deoxycytidine Quantitation), Cell Biolabs
Supplier: Cell Biolabs
STA-380 STA-380-5
102965-050EA 3015.38 USD InStock
102965-050 102965-048
Global DNA Methylation ELISA Kit (5’-methyl-2’-deoxycytidine Quantitation), Cell Biolabs
Assays ELISAs
The Global DNA Methylation ELISA Kit is a competitive ELISA for the quantitative measurement of 5-methylcytosine (5MedCyd)
The unknown 5MedCyd samples or 5MedCyd standards are first added to a 5MedCyd DNA conjugate coated EIA plate. After a brief incubation, an anti-5MedCyd monoclonal antibody is added, followed by an HRP conjugated secondary antibody. The 5MedCyd content in unknown samples is determined by comparison with a predetermined 5MedCyd standard curve. This kit is suitable for use with genomic DNA from cells or tissues, and can detect as little as 150 nM of 5MedCyd.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty