Would you like to log in again?
OxiSelect™ Human Oxidized LDL ELISA Kits, Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™
STA-389 STA-358 STA-369 STA-388
102965-064EA 910.77 USD InStock
102965-064 102965-040 102965-066 102965-000
OxiSelect™ Human Oxidized LDL ELISA Kits, Cell Biolabs
Assays ELISAs
OxiSelect™ Human Oxidized LDL ELISA Kits provide a convenient method to measure one of the markers of Oxidized LDL (OxLDL) in human serum or plasma samples
Kits are available to measure one of three markers of oxidation on the protein portion of LDL: MDA-LDL, CML-LDL, or HNE-LDL. In addition, the OxPL kit measures oxidation of the phospholipid portion of LDL. The kits contain a copper oxidized LDL as a standard against which unknown samples may be measured.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty