Would you like to log in again?
Serum Triglyceride Quantification Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-396 STA-397
102965-080EA 658.46 USD InStock
102965-080 102965-078
Serum Triglyceride Quantification Kits, Cell Biolabs
Assays Cellular Assays
Cell Biolabs’ Serum Triglyceride Quantification Kits measure triglyceride concentrations in serum, plasma, and lysates by a coupled enzymatic reaction system.  First, lipase hydrolyzes the triglyceride ester bond, yielding glycerol.  The free glycerol is then phosphorylated and oxidized, producing hydrogen peroxide which reacts with the kit’s probe
Kits are available with either colorimetric or fluorometric detection in a 96-well plate reader.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty