Would you like to log in again?
Ras Activation Assays, Cell Biolabs
Supplier: Cell Biolabs
STA-400-N STA-400-K STA-400 STA-400-H
102965-098EA 640 USD InStock
102965-098 102965-086 102965-094 102965-090
Ras Activation Assays, Cell Biolabs
Assays Protein Assays
Cell Biolabs' Ras Activation Assays use visible agarose beads to selectively precipitate the active form of specific Ras protein of interest
Assays are available to detect specific isoforms H-Ras, K-Ras, and N-Ras, as well as a Pan-Ras assay that detects all three isoforms. The kits use a safe, non-radioactive format.
The precipitated small GTPase is then detected by Western blot using a target-specific antibody included in the kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty