Would you like to log in again?
HDL Purification Kit (Ultracentrifugation Free), Cell Biolabs
Supplier: Cell Biolabs
STA-607
102965-214EA 473.85 USD InStock
102965-214
HDL Purification Kit (Ultracentrifugation Free), Cell Biolabs
Protein Purification Protein Purification Kits
Traditional ultracentrifugation methods for purifying LDL, VLDL or HDL can be time consuming and inconvenient
Our HDL Purification Kit uses dextran sulfate to selectively and separately purify HDL fractions for a variety of downstream applications. No ultracentrifuge is necessary, and purification steps use lower centrifugation speeds available with most benchtop centrifuges. Each prep purifies up to 10 mL of plasma or serum, and the average yield is approximately 4.5 mg/mL of HDL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty