Would you like to log in again?
Human Cholesteryl Ester Transfer Protein (CETP) ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-614
102965-230EA 763.08 USD InStock
102965-230
Human Cholesteryl Ester Transfer Protein (CETP) ELISA Kit, Cell Biolabs
Assays ELISAs
Cell Biolabs’ CETP ELISA Kit is an enzyme immunoassay developed for the detection and quantitation of Cholesteryl Ester Transfer Protein in human plasma, serum or other biological fluid samples
The kit detects CETP from human samples and has a detection sensitivity limit of 60 ng/mL CETP. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and unknown samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty