Would you like to log in again?
OxiSelect™ Glutathione Reductase Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-812
102965-280EA 443.08 USD InStock
102965-280
OxiSelect™ Glutathione Reductase Assay Kit, Cell Biolabs
Assays Enzyme Assays
OxiSelect™ Glutathione Reductase Assay Kit is a quantitative assay for measuring glutathione reductase activity within plasma, erythrocytes, tissues, and cell lysates
Glutathione reductase activity is defined as 1 unit of enzyme reducing 1 µmole oxidized glutathione (GSSG) per minute at pH 7.6 and 25ºC.  The kit employs a simple enzymatic recycling reaction for glutathione quantification where the reduction of a chromagen is correlated to glutathione reductase enzymatic activity. The kit can detect glutathione reductase activity as low as 0.6 mU/mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty