Would you like to log in again?
OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-859
102965-322EA 566.15 USD InStock
102965-322
OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit, Cell Biolabs
Assays Cellular Assays
OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit is a quantitative assay for measuring the antioxidant potential within various samples
Following the reduction of ferric iron (Fe3+) to ferrous iron (Fe2+) by antioxidants present in the sample, the kit colorimetric probe develops a blue color that is read colorimetrically at 540-600nm. The antioxidant potential of samples is determined based on an iron standard curve and results are calculated at Fe2+ equivalents (µM) or FRAP value. This kit provides fast results after a quick 10 minute protocol, and it is suitable for use with serum, plasma, lysates, biological fluids, and purified food or drug extracts.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty