Would you like to log in again?
QuickTiter™ Adenovirus Quantitation Kit, Cell Biolabs
Supplier: Cell Biolabs
QuickTiter™
VPK-106
102965-342EA 787.69 USD InStock
102965-342
QuickTiter™ Adenovirus Quantitation Kit, Cell Biolabs
Assays Cellular Assays
QuickTiter™ Adenovirus Quantitation Kit provides a quick method for measuring the viral nucleic acid content of your adenovirus prep
This assay may be performed either before or after purification of your virus. The kit uses an ultra-fast 45-60 minute procedure to measure the viral nucleic acid content of purified adenovirus or unpurified supernatant. The limit of detection is 10^10 viral particles/mL from 100 µL of adenoviral supernatant.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty