Would you like to log in again?
QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated p24 ELISA), Cell Biolabs
Supplier: Cell Biolabs
QuickTiter™
VPK-107-T VPK-107 VPK-107-5
102970-592EA 849.23 USD InStock
102970-592 102965-346 102965-344
QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated p24 ELISA), Cell Biolabs
Assays ELISAs
The QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated HIV p24) provides a unique and more accurate way to measure physical lentiviral titer
This kit substantially minimizes the problem of overestimating lentiviral titer with traditional p24 ELISA kits. The traditional p24 ELISA detects both virus-associated p24 and free p24 generated by 293 cells during transient transfection. Free p24 can account for a substantial portion of total p24 in the supernatant. Therefore, the ELISA typically overestimates the quantity of lentivirus present
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty