Would you like to log in again?
OxiSelect™ Intracellular ROS Assay Kit (Green Fluorescence), Cell Biolabs
Supplier: Cell Biolabs
STA-342 STA-342-5 STA-342-T
102965-786EA 363.08 USD InStock
102965-786 102965-784 102965-782
OxiSelect™ Intracellular ROS Assay Kit (Green Fluorescence), Cell Biolabs
Assays Cellular Assays
The OxiSelect™ ROS Assay Kit is a cell-based assay for measuring hydroxyl, peroxyl, and other reactive oxygen species activity within a cell
The assay employs the cell-permeable fluorogenic probe DCFH-DA, which diffuses into cells and is deacetylcated by cellular esterases into the non-fluorescent DCFH. In the presence of ROS, DCFH is rapidly oxidized to highly fluorescent DCF. After a quick one hour protocol, fluorescence is read on a standard fluorometric plate reader. The kit is highly sensitive to as low as 10 pM of DCF.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty