Would you like to log in again?
Small GTPase Agarose Beads, Cell Biolabs
Supplier: Cell Biolabs
STA-433 STA-432 STA-420 STA-412 STA-418 STA-421 STA-431 STA-410 STA-411 STA-419
102965-818EA 627.69 USD InStock
102965-818 102965-162 102966-240 102965-160 102966-238 102965-822 102965-820 102965-158 102965-156 102966-244
Small GTPase Agarose Beads, Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Small GTPase Assay Beads are agarose beads containing a binding domain that selectively precipitates the active form of the small GTPase target of interest
The beads contain a light red color to allow easy visualization during the pulldown process. These are the same agarose beads found in Cell Biolabs' Small GTPase Activation Assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty