Would you like to log in again?
Lactose Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
MET-5001
102966-204EA 467.69 USD InStock
102966-204
Lactose Assay Kit, Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Lactose Assay Kit measures total lactose in milk based food products or biological samples (such as blood or urine from lactating mammals)
Lactose is first cleaved into glucose and galactose by lactase. Glucose is then oxidized by glucose oxidase, yielding hydrogen peroxide and D-gluconic acid. The hydrogen peroxide released from this reaction is detected by a fluorometric probe in a 1:1 ratio. Lactose levels in unknown samples are determined based on a lactose standard curve. The Lactose Assay Kit can detect lactose concentrations as low as 10µM.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty