Would you like to log in again?
OxiSelect™ Cellular UV-Induced DNA Damage Staining Kit (CPD), Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™
STA-327
102966-234EA 763.08 USD InStock
102966-234
OxiSelect™ Cellular UV-Induced DNA Damage Staining Kit (CPD), Cell Biolabs
Assays Cellular Assays
OxiSelect™ Cellular UV-Induced DNA Damage Staining Kit measures the formation of cyclobutane pyrimidine dimers (CPD) by immunofluorescence
Cells are first seeded in a 96-well tissue culture plate. Wells are then UV irradiated to induce DNA damage. After fixation and denaturation, cells containing the DNA lesions are probed with an anti-CPD antibody, followed by a FITC conjugated secondary antibody. The unbound secondary antibody is removed during a wash step, and stained cells can then be visualized with a fluorescence microscope.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty