Would you like to log in again?
QuickTiter™ HCV Core Antigen ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
QuickTiter™
VPK-151
102966-342EA 1076.92 USD InStock
102966-342
QuickTiter™ HCV Core Antigen ELISA Kit, Cell Biolabs
Assays ELISAs
Cell Biolabs’ QuickTiter™ HCV Core Antigen ELISA Kit is an enzyme immunoassay developed for detection and quantitation of the HCV core protein
The kit has detection sensitivity limit of 1 ng /mL HCVcAg. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and HCV samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty