Would you like to log in again?
CytoSelect™ Transmigration Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
CBA-216 CBA-212
102963-832EA 855.38 USD InStock
102963-832 102966-500
CytoSelect™ Transmigration Assay Kits, Cell Biolabs
Assays Cellular Assays
Cell transmigration involves the migration of cells across the endothelium
CytoSelect™ Transmigration Assays measure the interactions with, and transmigration of, the endothelium by leukocytes or tumor cells. The assays use a Boyden Chamber in which endothelial cells may be seeded to form a monolayer prior to adding the migratory cells of interest. Migratory cells move through the endothelial layer and are counted using the fluorescent dye provided in each kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty