Would you like to log in again?
4-Chip Disposable Hemocytometer, Neubauer Improved, Bulldog Bio
Supplier: Bulldog Bio
DHC-N404 DHC-N420 DHC-N408
102966-652EA 46.15 USD InStock
102966-652 102966-632 102966-650
4-Chip Disposable Hemocytometer, Neubauer Improved, Bulldog Bio
Slides Counting Chambers
An improvement on an old favorite; the 4-Chip has twice as many tests as the C-Chip for a comparable price, and the same ease of use.
  • Four tests per chip for high throughput
  • Highly repeatable counts
  • Integrated coverslip for ease of use
  • Completely disposable
The 4-Chip is a new style of disposable plastic hemocytometer that improves upon the popular C-Chip design
With a conventional glass hemocytometer, improper fit between coverslip and slide can change the volume of the sample in the chamber, causing variability in counts. Moreover, these “unretained” samples may lead to cross-contamination or far worse, exposing researchers to infectious agents. The 4-Chip’s all-in-one chamber with an integrated coverslip solves this problem. The 4-Chip contains four precision-engineered, 100µm height counting chambers that do not need coverslips. Each chamber has a micro-etched Neubauer Improved grid. Simply pipette 6µL of sample into one of the four chambers, place on the microscope, and it is ready to count. No more tiresome washing either, simply throw it away when finished. The 4-Chip is that simple.
With twice as many tests at almost the same price as the 2-chamber version, this product effectively cuts the lab’s costs by nearly half.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty