Would you like to log in again?
NEB PCR Cloning Kit (Without Competent Cells), New England Biolabs
Supplier: New England Biolabs
E1203S E1203S
102969-096EA 188 USD InStock
102969-096 CA102969-096
NEB PCR Cloning Kit (Without Competent Cells), New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
This PCR Cloning Kit contains an optimized Cloning Mix containing a proprietary ligation enhancer and a linearized vector that uses a novel mechanism for background colony suppression to give a low background.
It allows simple and quick cloning of any PCR amplicon, whether the amplification reactions are performed with proofreading DNA polymerases, such as Q5® which produce blunt ends; or nonproofreading DNA polymerases, such as Taq or Taq mixes (OneTaq®, LongAmp® Taq) which produce single base overhangs. This is possible due to “invisible” end polishing components in the master mix that are active during the ligation step only if needed. The kit also allows direct cloning from amplification reactions without purification, and works well whether or not the primers used in the PCR possess 5´-phosphate groups.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty