Would you like to log in again?
NEBNext Ultra II Ligation Module, New England Biolabs
Supplier: New England Biolabs
E7595L E7595S E7595S E7595L
102969-106EA 415 USD InStock
CA102969-104 102969-106 CA102969-106 102969-104
NEBNext Ultra II Ligation Module, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
The NEBNext Ultra II Ligation Module has been optimized to efficiently ligate DNA adaptors compatible with Illumina® sequencing to end-repaired, dA-tailed DNA fragments
The module is optimized for use with the NEBNext Ultra II End Repair/dA-Tailing Module (NEB #E7546), and is part of the Ultra II workflow which enables high yield preparation of high quality libraries from 500 picograms to 1 microgram of input DNA. It is also designed for use with NEBNext Ultra II Q5® Master Mix (NEB #M0544) and NEBNext Singleplex or Multiplex Oligos for Illumina (NEB #E7350, #E7335, #E7500, #E7600 or #E7535).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty