Would you like to log in again?
CAPILLARY GLASS WITH FILAMENT PK 225
Supplier: HARVARD APPARATUS MS
Glass^ Premium Standard Wall Borosilicate Capillary Glass with Fillament, Model G150F-4, OD 1.5 mm, ID 0.86 mm, Ends are fire polished to prevent damage to the rubber gaskets when inserted into electrode holders, Glass is also cleaned with deionized water before being packed in dust-free containers
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPILLARY GLASS WITH FILAMENT PK 225 102970-580 640793 Pack of 225 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty