Would you like to log in again?
L-Glutamine LifeFactor, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1013
102970-732EA 69.85 USD InStock
102970-732
L-Glutamine LifeFactor, Lifeline Cell Technology
Cell Culture Supplements and Additives
L-Glutamine LifeFactor is used with basal medium and human cells.
  • Used with human endothelial cells
  • Used with human smooth muscle cells
  • Used with basal medium
L-Glutamine LifeFactor -- 25 mL may be used with: Lifeline® VascuLife® basal medium and human smooth muscle cells, or Lifeline® VascuLife® basal medium and human endothelial cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty