Would you like to log in again?
FibroLife® S2 Fibroblast Medium Kit, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0011 LS-1038
102970-816EA 195.85 USD InStock
102970-816 102970-854
FibroLife® S2 Fibroblast Medium Kit, Lifeline Cell Technology
Cell Culture Supplements and Additives
FibroLife® S2 Medium Complete Kit is optimized for low serum (2%) culture of fibroblasts. FibroLife® S2 supports the growth of these cells in a 2% serum environment at rates equal to or greater than comparable media supplemented with up to 10% FBS.
  • Optimized for low serum culture of fibroblasts
  • Contains no phenol red or antimicrobials to affect cell performance.
FibroLife® S2 Medium Complete Kit is optimized for low serum (2%) culture of fibroblasts
The Complete Kit includes FibroLife® Basal Medium and the FibroLife® S2 LifeFactors® Kit (containing the associated supplements and growth factors) to allow you to prepare fresh medium. FibroLife® contains no phenol red or antimicrobials to affect cell performance.
FibroLife® Complete Kits contain FibroLife® Basal Medium and the associated LifeFactors® Kit. FibroLife® Basal Medium requires the associated LifeFactors®.
FibroLife® S2 LifeFactors® Kit includes: rh FGF basic 0.5 mL, L-Glutamine 18.75 mL, Hydrocortisone Hemisuccinate 0.5 mL, Ascorbic Acid 0.5 mL, rh Insulin 0.5 mL, FBS 10 mL, Gentamicin and Amphotericin B 0.5 mL.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty