Would you like to log in again?
Extract-P LifeFactor, Bovine, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1037
102970-850EA 114.62 USD InStock
102970-850
Extract-P LifeFactor, Bovine, Lifeline Cell Technology
Cell Culture Supplements and Additives
Extract P may be used with basal medium, and human cells and keratinocytes.
  • Used with basal medium
  • Used with human reproductive cells
  • Used with human corneal epithelial cells
  • Used with human airway epithelial cells
  • Used with human mammary epithelial cells
  • Used with human bladder apex epithelial cells
Extract P may be used with these Lifeline® culture media and human cell types:
ProstaLife™ basal medium and human reproductive epithelial cells, DermaLife basal medium or DermaLife calcium-free basal medium and human keratinocytes, OcuLife™ basal medium and human corneal epithelial cells, UroLife™ A basal medium and human bladder apex epithelial cells, BronchiaLife™ basal medium and human airway epithelial cells, MammaryLife™ basal medium and human mammary epithelial cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty