Would you like to log in again?
T3 Triiodo-l-Thyronine LifeFactor, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1044
102970-852EA 67.54 USD InStock
102970-852
T3 Triiodo-l-Thyronine LifeFactor, Lifeline Cell Technology
Cell Culture Supplements and Additives
T3 triiodo-l-thyronine is a thyroid hormone that regulates cell differentiation and protein expression.
  • Thyroid hormone
  • Regulates cell differentiation
  • Regulates protein expression
T3 triiodo-l-thyronine is a thyroid hormone that regulates cell differentiation and protein expression.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty