Would you like to log in again?
Trypsin
Supplier: Lifeline Cell Technology
CM-0046
102970-884EA 67.69 USD InStock
102970-884
Trypsin
Enzymes
Trypsin acts as an enzyme to help dissociate cells from their culture vessel.
  • Helps prevents clumping of cells
  • Calcium chelator
  • Trypsin 0.05% EDTA 0.02%
  • Xeno-free
Trypsin acts as an enzyme to help dissociate cells from their culture vessel. EDTA is a calcium chelator that helps to prevent clumping of cells. The Xeno-Free version contains no non-human animal-derived components.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty