Would you like to log in again?
Trypsin Neutralizing Solution, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
CM-0018 CM-0047
102970-808EA 48.46 USD InStock
102970-808 102970-886
Trypsin Neutralizing Solution, Lifeline Cell Technology
Cell Culture Supplements and Additives
Trypsin acts as an enzyme to help dissociate cells from their culture vessel.
  • Acts as an enzyme to break down proteins
Trypsin acts as an enzyme to break down proteins to dissociate cells from the vessel in which they're being cultured. Trypsin Neutralizing Solution Xeno-Free contains no non-human animal-derived component.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty