Would you like to log in again?
NewBlot™ Stripping Buffers
Supplier: LI-COR®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
928-40032 928-40028 928-40030
102673-518EA 201.85 USD InStock
102673-518 102971-254 102673-520
NewBlot™ Stripping Buffers
Buffers Electrophoresis Buffers Stripping Buffers
NewBlot Stripping Buffers are optimized for near-infrared (NIR) Western blots, including blots detected with IRDye® secondary antibodies
  • Ideal for removal of secondary antibody after detection of highly abundant targets. See performance table for stripping buffer comparisons.
  • Optimized for stripping NIR Western blots detected with IRDye 800CW, IRDye 680LT, or IRDye 680RD secondary antibodies.
  • NewBlot PVDF Buffer should not be used with IRDye 700DX conjugates.
Stripping and reprobing a Western blot is a common laboratory method. Many labs choose to make their own stripping buffer. What else could you be doing with that time?
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty