Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Monarch® DNA Cleanup Kits, New England Biolabs

Supplier: New England Biolabs
Monarch®
T1031L T1032L T1030L T1030S T1021L T1034L T1020S T1030L T1021L T1032L T1020L T1034L T1030S T1016L T1020S T1016L T1020L T1031L
102971-682EA 43 USD InStock
102971-682 102971-670 FPT1031L 102971-680 102971-674 102971-672 102971-668 102971-678 102971-688 102971-676 CA102971-672 CA102971-682 CA102971-670 CA102971-688 CA102971-676 CA102971-674 CA102971-668 CA102971-678
Monarch® DNA Cleanup Kits, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
Monarch DNA Cleanup Kits, available for both gel extraction and reaction cleanup, are optimized for maximum performance and minimal environmental impact.

  • Elute in as little as 6 μl
  • Prevent buffer retention and salt carry-over with optimized column design
  • Save time with fast, user-friendly protocols
  • No need to monitor pH or add isopropanol
  • Buffers and columns available separately
  • Significantly less plastic used when compared with other kits
  • Responsibly-sourced and recyclable packaging

These kits rapidly and reliably purify up to 5 μg of concentrated, highquality DNA and utilize a bind/wash/elute workflow with minimal incubation and spin times. The columns provided with each kit are specifically designed to eliminate buffer retention and the risk of carryover of contaminants, enabling elution of sample in volumes as low as 6 μl. Optimized buffer systems eliminate the need for monitoring and adjusting pH

The Monarch DNA Gel Extraction Kit can be used to quickly purify DNA from agarose gels. Unlike other kits, there is no need to add isopropanol to the melted agarose prior to loading on the column, saving you a step.
The Monarch PCR & DNA Cleanup Kit (5 μg) can be used to purify DNA from a variety of enzymatic reactions. The DNA Wash Buffer provided ensures enzymes, short primers (≤ 25 nt), detergents and other low-molecular weight reaction components are removed.
Both kits use significantly less plastic and responsibly-sourced, recyclable packaging, so you can feel good about choosing Monarch for your DNA purification.

Specifications
• Binding Capacity: up to 5 μg
• DNA Size Range: ~50 bp to 25 kb
• Typical Recovery: DNA (50 bp to 10 kb): 70–90% DNA (11–25 kb): 50–70%
• Elution Volume: ≥ 6 μl
• Purity: A260/A280 ≥ 1.8
• Protocol Time: Gel Extraction: 10 min of spin and incubation time PCR & DNA Cleanup: 5 min of spin and incubation time
• Compatible Downstream Applications: ligation, restriction digestion, labeling and other enzymatic manipulations, library construction and DNA sequencing

Caution: The kit should be stored at room temperature. Always keep buffer bottles tightly closed and keep columns sealed in the enclosed zip-lock bag. For information regarding the composition of buffers, please consult the Safety Data Sheets. Proper laboratory safety practices should be employed, including the use of lab coats, gloves and eye protection.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty