Would you like to log in again?
Monarch™ Plasmid Miniprep Kit and Components, New England Biolabs®
Supplier: New England Biolabs
Monarch®
T1015L T1010L T1017L T1010S T1013L T1015L T1011L T1013L T1017L T1010L T1012L T1014L T1010S T1011L
102971-692EA 43 USD InStock
102971-692 102971-690 102971-686 102971-696 102971-684 102971-694 102971-666 102971-698 CA102971-690 CA102971-694 CA102971-666 CA102971-698 CA102971-686 CA102971-696
Monarch™ Plasmid Miniprep Kit and Components, New England Biolabs®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Monarch Plasmid Miniprep Kit is a rapid and reliable method for the purification of high quality plasmid DNA.
  • Elute in as little as 30 μl
  • Eliminate buffer retention and salt carryover with optimized column design
  • Save time with fast, user-friendly protocols
  • Easily label columns using tab and frosted surfaces
  • Monitor completion of certain steps and easily identify buffers with colored buffer system
  • No need to add RNase before starting
  • Reduced environmental impact
  • Recyclable packaging
This method employs standard cell resuspension, alkaline lysis, and neutralization steps, with the additional benefit of color indicators at certain steps to easily monitor completion. Unique wash buffers ensure salts, proteins, RNA and other cellular components are removed, allowing low-volume elution of concentrated, highly pure DNA. Protocols are fast and user friendly, saving you valuable time. Elution in as little as 30 μl provides concentrated DNA for use in downstream applications, such as restriction digests, DNA sequencing, PCR and other enzymatic manipulations. Designed with sustainability in mind, Monarch kits use significantly less plastic and responsibly-sourced, recyclable packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty