Would you like to log in again?
C-peptide Chemiluminescence ELISA
Supplier: ALPCO
80-CPTHU-CH01
102977-818EA 746.15 USD InStock
102977-818
C-peptide Chemiluminescence ELISA
Assays ELISAs
The ALPCO Chemi Human C-peptide ELISA is an FDA registered for In Vitro diagnostic use tool for the quantification of human C-peptide in serum and plasma samples in both clinical and research laboratory settings.
  • Broad dynamic range of 4.5 to 12960 pg/ml
  • Functional sensitivity: <18 pg/ml
  • LOD: <4.32 pg/ml
Measurement of C-peptide, a 31 amino acid peptide, is being used to further understand diabetes mellitus, hypoglycemia and insulinoma. C-peptide is a byproduct formed in the process involving a series of enzymatic cleavages of preproinsulin and proinsulin, with proinsulin being the immediate precursor of insulin and C-peptide. C-peptide and insulin are known to be released in equimolar amounts from beta cells of pancreas. Since the half-life of C-peptide is about 30 minutes compared to insulin which is only about 3 minutes, measuring C-peptide may be more attractive for indirectly estimating glucose stimulated insulin secretion, understanding beta cell function and/or identifying beta cell functional mass.
The STELLUX® Chemi Human C-Peptide ELISA is a FDA Class I in vitro diagnostic tool for the quantification of human C-peptide in a clinical setting or research laboratory. Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, a single STELLUX® Chemi Human C-peptide ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 36 samples in duplicate. The shelf-life of the components of the STELLUX® Chemi Human C-Peptide ELISA and the resealable microplate pouch allow for convenient storage for future use if the entire kit is not needed at one time.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty