Would you like to log in again?
C-peptide ELISA Jumbo (10-Plate)
Supplier: ALPCO
80-CPTHU-E10
102977-820EA 4535.38 USD InStock
102977-820
C-peptide ELISA Jumbo (10-Plate)
Assays ELISAs
The C-peptide ELISA tool for the quantification of human C-peptide in serum and plasma samples in both clinical and research laboratory settings.
Certifications: FDA Class I Device.
Caution: For In Vitro diagnostic use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty