Would you like to log in again?
Insulin Chemiluminescence ELISA Jumbo (10-pack)
Supplier: ALPCO
80-INSHU-CH10
102977-822EA 5030.77 USD InStock
102977-822
Insulin Chemiluminescence ELISA Jumbo (10-pack)
Assays ELISAs
The chemi human insulin ELISA jumbo (10-plate) measures the concentration of insulin in human serum, heparin plasma, and tissue culture supernatants. Also available in single plate format.
  • Broad dynamic range of 5 to 30000 pg/ml
  • Superior sensitivity compared other products on the market
  • LOD: <2.0 pg/ml
  • LLOQ: 20 pg/ml
The STELLUX® chemi human insulin ELISA measures the concentration of insulin protein products and offers superior performance compared to other similar kits in the market. The assay utilizes a dual-monoclonal antibody sandwich format, tri-level control set, and 96-well microplate comprising removable strips. The assay can measure up to 37 samples in duplicate and can be used in a variety of sample types.
Broad dynamic range (5 to 30000 pg/ml) - significantly reduces sample preparation time by eliminating dilution steps and the guesswork involved in determining dilution factors, saving time and money. No guessing, no diluting, no repeating. Sensitivity - superior assay sensitivity compared to current competitive products on the market. LOD: <2.0 pg/ml, with a lower limit of quantification of 20 pg/ml.
One assay to fit all sample types - the assay works with human serum, heparin plasma and tissue culture supernatants.
Certifications: FDA Class I device. For in vitro diagnostic use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty