Would you like to log in again?
Ultrasensitive Insulin ELISA Jumbo (10-plate)
Supplier: ALPCO
80-INSHUU-E10
102977-826EA 4024.62 USD InStock
102977-826
Ultrasensitive Insulin ELISA Jumbo (10-plate)
Assays ELISAs
The ultrasensitive insulin ELISA jumbo (10-plate) kit quantifies the concentration of insulin protein in human serum or plasma samples.
  • Only 25 µl of sample is sufficient for the assay
  • Superior sensitivity of 0.135 µIU/ml
  • Dynamic range of 0.15 to 20 µIU/ml
The ultrasensitive insulin ELISA kit is a FDA registered in vitro diagnostic tool for the quantification of human insulin in a clinical setting or research laboratory. The Ultrasensitive Insulin ELISA utilizes a dual-monoclonal antibody sandwich ELISA format, diabetes control, and 96-well microplate comprising removable strips. The Ultrasensitive Insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if the entire kit is not needed at one time.
Certifications: FDA Class I device. For in vitro diagnostic use.
Packaging: Single kit packaging is also available.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty