Would you like to log in again?
STELLUX® Chemi Rodent Insulin ELISA Jumbo, ALPCO
Supplier: ALPCO
80-INSMR-CH10
102977-828EA 5030.77 USD InStock
102977-828
STELLUX® Chemi Rodent Insulin ELISA Jumbo, ALPCO
Assays ELISAs
The STELLUX® chemi rodent insulin ELISA jumbo measures the concentration of insulin protein products in mouse I, mouse II, rat I and rat II proinsulin genes.
  • Low sample volume of 5 µl
Can measure up to 370 samples in duplicate.
The STELLUX® chemi rodent insulin ELISA Jumbo utilizes a dual-monoclonal antibody sandwich format, tri-level control set, and contains 10 plates comprising removable strips.
Caution: Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty