Would you like to log in again?
IdeZ Protease (IgG-specific)
Supplier: New England Biolabs
P0770S P0770S
102978-556EA 487 USD InStock
102978-556 CA102978-556
IdeZ Protease (IgG-specific)
Enzymes
IdeZ Protease (IgG-specific) is a recombinant antibody specific protease cloned from Streptococcus equi subspecies zooepidemicus that recognizes all human, sheep, monkey, and rabbit IgG subclasses, specifically cleaving at a single recognition site below the hinge region, yielding a homogenous pool of F(ab´)2 and Fc fragments
IdeZ Protease more effectively cleaves murine IgG2a than IdeS.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty