Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KillerTRAIL™ Dilution & Storage Buffer, AdipoGen Life Sciences

Supplier: AdipoGen Life Sciences
AG-10T-0001-R500
102978-722EA 110.77 USD InStock
102978-722
KillerTRAIL™ Dilution & Storage Buffer, AdipoGen Life Sciences
Buffers Biological Buffers
Dilution & Storage Buffer

Formulation: Liquid. 20mM HEPES, pH 7.4, containing 300mM NaCl, 0.01% Tween 20, 1% sucrose and 1mM DTT.

Endotoxin content: <0.01EU/µl purified protein (LAL test; Lonza).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty