Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PBS Endotoxin-free (sterile), AdipoGen Life Sciences

Supplier: AdipoGen Life Sciences
IAX-900-001-L010 IAX-900-001-LD15 IAX-900-001-3010
102979-260EA 34.62 USD InStock
102979-260 102979-262 102979-264
PBS Endotoxin-free (sterile), AdipoGen Life Sciences
Buffers Cell Culture Buffers
Endotoxin free buffer used for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g

Formulation: Liquid. 1.5ml (IAX-900-001-LD15) or 10ml (IAX-900-001-L010) PBS pH 7.2 in sterile filtered double-distilled water (ddWater), without any additives.

Endotoxin: <0.0002EU/µg, after repeated purification cycles over activated charcoal.

ODNs).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty