Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Phosphoprotein Associated with Glycosphingolipid-enriched Microdomains 1;Csk-binding Protein Rabbit Polyclonal Antibody

Supplier: AdipoGen Life Sciences
AG-25T-0108-C100
102979-976EA 685.38 USD InStock
102979-976
Anti-Phosphoprotein Associated with Glycosphingolipid-enriched Microdomains 1;Csk-binding Protein Rabbit Polyclonal Antibody
Antibodies
PAG is a ubiquitously expressed transmembrane adapter protein. Depending on its phosphorylation status, it appears as diffuse band(s) of 80-90kDa by SDS-PAGE, with phosphorylation leading to a shift towards a higher apparent molecular mass. Tyrosine phosphorylated PAG recruits Csk to the membrane and negatively regulates Src family kinases via Csk-mediated phosphorylation. Upon T cell receptor engagement, PAG becomes dephosphorylated, thereby displacing Csk from the membrane and enabling the activation of the Src kinases Fyn and Lck. PAG negatively regulates TCR (T cell antigen receptor)-mediated signaling in T cells and FCER1 (high affinity immunoglobulin epsilon receptor)-mediated signaling in mast cells. It promotes CSK activation and recruitment to lipid rafts, which results in LCK inhibition. It inhibits immunological synapse formation by preventing dynamic arrangement of lipid raft proteins and may be involved in cell adhesion signaling.

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty