Would you like to log in again?
Human NMNAT1 (from E. coli), AdipoGen Life Sciences
Supplier: AdipoGen Life Sciences
AG-40T-0092-C010 AG-40T-0092-3050 AG-40T-0092-C050
102980-428PK 969.23 USD InStock
102980-428 102980-426 102980-424
Human NMNAT1 (from E. coli), AdipoGen Life Sciences
Enzymes
Nuclear NMNAT isoform
  • UniProt link Q9HAN9: NMNAT1 (human)
Catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. It can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. Interacts with PARP-1/ARTD1. Protects against axonal degeneration following mechanical or toxic insults. Widely expressed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty