Would you like to log in again?
Human NMNAT3 (from E. coli), AdipoGen Life Sciences
Supplier: AdipoGen Life Sciences
AG-40T-0093-C010 AG-40T-0093-C050 AG-40T-0093-3050
102980-430EA 290.77 USD InStock
102980-430 102980-434 102980-432
Human NMNAT3 (from E. coli), AdipoGen Life Sciences
Enzymes
Mitochondrial NMNAT isoform
  • UniProt link Q96T66: NMNAT3 (human)
Catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. Can use tiazofurin monophosphate as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD+. For the pyrophosphorolytic activity, mitochondrial NMNAT isoform can use NAD(+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP+, NADPH and NAADP+. Protects against axonal degeneration following injury.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty