Would you like to log in again?
TDO (from Escherichia coli), AdipoGen Life Sciences
Supplier: AdipoGen Life Sciences
AG-40T-0101-C050
102980-438EA 540 USD InStock
102980-438
TDO (from Escherichia coli), AdipoGen Life Sciences
Enzymes
Tryptophan 2,3-dioxygenase (TDO) is a heme-containing dioxygenase catalyzing the addition of molecular oxygen across the 2,3-double bond of the indole ring of tryptophan to form N-formylkynurenine (NFK)
  • UniProt link O77457: TDO (Anopheles gambiae)
In Anopheles gambiae, TDO is the only enzyme able to catalyze the first and rate-limiting step in L-Trp catabolism through the kynurenine pathway. Tryptophan is an essential amino acid for protein synthesis and also the precursor for production of a number of neurotransmitters, such as serotonin and melatonin; in mosquitoes, the kynurenine pathway is essential for eye pigmentation. Conceivably, the tryptophan-using pathways should be regulated in a coordinated manner in mosquitoes as well as in other species and TDO activation/inactivation processes could play an essential role in these phenomena.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty