Would you like to log in again?
Human Kynurenine Aminotransferase II (from E. coli)
Supplier: AdipoGen Life Sciences
AG-40T-0102-C050
102980-440EA 540 USD InStock
102980-440
Human Kynurenine Aminotransferase II (from E. coli)
Enzymes
Kynurenine aminotransferases (KATs) are pyridoxal-5’-phosphate-dependent enzymes that catalyze the conversion of L-kynurenine into kynurenic acid, a neuroactive metabolite whose unbalancing is associated with a number of brain disorders
  • NCBI link NP_872603: AADAT (human)
Biochemical and structural investigations revealed that L-kynurenine (L-KYN) recognition by hKAT II is achieved by exploiting structural features that are peculiar of this isoform, thus offering the possibility to select/design inhibitor molecules specifically targeting hKAT II to be used as modulators of kynurenic acid synthesis in the CNS. hKAT II is one of the aminotransferases involved in the pyridoxal 5'-phosphate (PLP)-dependent irreversible transamination of L-kynurenine (L-KYN) to kynurenic acid (KYNA) in the central nervous system. When assayed in vitro the protein also displays beta-elimination activity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty