Would you like to log in again?
Zmp1 (from Escherichia coli)
Supplier: AdipoGen Life Sciences
AG-40T-0103-C050
102980-442EA 540 USD InStock
102980-442
Zmp1 (from Escherichia coli)
Enzymes
M13 endopeptidases are single-pass, transmembrane zinc-metallopeptidases responsible in human for the regulation of the biological activity of many hormones and peptides and are involved in many important processes such as blood pressure regulation (Neprilysin or NEP1), cardiovascular development (Endothelin Converting Enzyme-1 or ECE-12), prevention of haemolytic reaction (KELL) and phosphate homeostasis (PHEX)
  • UniProt link O53649: Zmp1 (M. tuberculosis)
Zmp1 is a soluble enzyme member of the M13 endopeptidases family that plays a key role in Mycobacterium tuberculosis pathogenicity with a still unknown mechanism. Recently, it was shown to suppress inflammasome activation by inhibiting caspase-1 activation, thus preventing processing of pro-IL-1beta into IL-1beta and the consequent phagosome maturation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty