Would you like to log in again?
NAD Synthetase (from E. coli)
Supplier: AdipoGen Life Sciences
AG-40T-0108-C050
102980-452EA 540 USD InStock
102980-452
NAD Synthetase (from E. coli)
Enzymes
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways
  • UniProt link P0A5L6: NAD Synthetase (M. tuberculosis)
Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It can use both glutamine or ammonia as a nitrogen source. Catalytic activity: ATP + deamido-NAD+ + L-glutamine + H2O = AMP + diphosphate + NAD+ + L-glutamate
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty