Would you like to log in again?
NAD Synthetase (from E. coli)
Supplier: AdipoGen Life Sciences
AG-40T-0109-C050
102980-454EA 540 USD InStock
102980-454
NAD Synthetase (from E. coli)
Enzymes
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways
  • UniProt link P08164: NAD Synthetase (B. subtilis)
Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It was shown to be induced by heat shock, salt stress, oxidative stress, glucose limitation and oxygen limitation and is involved in biological processes like stress response and sporulation resulting in formation of a cellular spore. Catalytic activity: ATP + deamido-NAD+ + NH3 = AMP + diphosphate + NAD+
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty